Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
G6PC
Entrez Gene ID
102170730
Gene Name
glucose-6-phosphatase catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.19: 42622247 - 42632786 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693877.3 XP_005693934.1 1 - 2 404 63
GenBank mRNA JO432881.2 - 1 - 2 444 63

View Full Product Details