Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEX11B
Entrez Gene ID
102184963
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
-
Chromosome Location
Chr.3: 99076746 - 99082382 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677737.3 XP_005677794.1 2 - 3 237 70
GenBank mRNA JO387383.2 - 2 - 3 283 70

View Full Product Details