Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB18
Entrez Gene ID
102186880
Gene Name
RAB18, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.13: 36386567 - 36414586 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287560.1 NP_001274489.1 2 - 3 176 101
GenBank mRNA JQ417406.1 - 2 - 3 176 101

View Full Product Details