Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED6
Entrez Gene ID
102172570
Gene Name
transmembrane p24 trafficking protein 6
Gene Aliases
-
Chromosome Location
Chr.18: 37753066 - 37759401 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692188.3 XP_005692245.1 3 - 4 866 109

View Full Product Details