Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC1A
Entrez Gene ID
102172976
Gene Name
C-type lectin domain family 1 member A
Gene Aliases
-
Chromosome Location
Chr.5: 98299798 - 98320304 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048491.1 XP_017903980.1 4 - 5 1145 104
XM_018048492.1 XP_017903981.1 4 - 5 500 104
GenBank mRNA JT757873.1 - 5 - 6 760 104

View Full Product Details