Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA7B
Entrez Gene ID
102173996
Gene Name
golgin A7 family member B
Gene Aliases
-
Chromosome Location
Chr.26: 32610682 - 32623915 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KA310371.1 - - 2179 78

View Full Product Details