Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRR2
Entrez Gene ID
102176240
Gene Name
gamma-aminobutyric acid type A receptor rho2 subunit
Gene Aliases
-
Chromosome Location
Chr.9: 48142689 - 48184445 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JT704336.1 - 3 - 4 179 86

View Full Product Details