Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB3B
Entrez Gene ID
102190011
Gene Name
RAB3B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.3: 26416254 - 26489570 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044093.1 XP_017899582.1 3 - 4 464 99
GenBank mRNA JT646517.1 - 4 - 5 422 99

View Full Product Details