Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED31
Entrez Gene ID
102191551
Gene Name
mediator complex subunit 31
Gene Aliases
-
Chromosome Location
Chr.19: 25043860 - 25050223 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693428.3 XP_005693485.1 2 - 3 203 110

View Full Product Details