Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM8A1
Entrez Gene ID
102172224
Gene Name
family with sequence similarity 8 member A1
Gene Aliases
-
Chromosome Location
Chr.23: 10536124 - 10547214 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038725.1 XP_017894214.1 2 - 3 947 111

View Full Product Details