Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGT8
Entrez Gene ID
102174638
Gene Name
UDP glycosyltransferase 8
Gene Aliases
-
Chromosome Location
Chr.6: 11268248 - 11381007 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049186.1 XP_017904675.1 3 - 4 1469 96

View Full Product Details