Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PPP1R11
- Entrez Gene ID
-
108633186
- Gene Name
- protein phosphatase 1 regulatory inhibitor subunit 11
- Gene Aliases
-
-
- Chromosome Location
-
Chr.23: 21155372 - 21158523 on Build ARS1
- Gene Symbol
- RNF39
- Entrez Gene ID
-
102171828
- Gene Name
- ring finger protein 39
- Gene Aliases
-
-
- Chromosome Location
-
Chr.23: 21158460 - 21163923 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KA305866.1 |
- |
4 - 5 |
3311 |
74 |
View Full Product Details