Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MINPP1
Entrez Gene ID
102187288
Gene Name
multiple inositol-polyphosphate phosphatase 1
Gene Aliases
-
Chromosome Location
Chr.26: 42272224 - 42305810 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041630.1 XP_017897119.1 2 - 3 1900 71

View Full Product Details