Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC8C
Entrez Gene ID
102173523
Gene Name
leucine rich repeat containing 8 VRAC subunit C
Gene Aliases
-
Chromosome Location
Chr.3: 66963367 - 67046194 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045728.1 XP_017901217.1 2 - 3 379 68
GenBank mRNA JT687017.1 - 2 - 3 2276 68
JT755413.1 - 6 - 7 2647 68

View Full Product Details