Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC16
Entrez Gene ID
102168272
Gene Name
tetratricopeptide repeat domain 16
Gene Aliases
-
Chromosome Location
Chr.11: 97943415 - 97957481 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055816.1 XP_017911305.1 3 - 4 287 86

View Full Product Details