Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBI
Entrez Gene ID
102182266
Gene Name
diazepam binding inhibitor, acyl-CoA binding protein
Gene Aliases
-
Chromosome Location
Chr.2: 64935651 - 64940897 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676252.3 XP_005676309.1 2 - 3 331 67
GenBank mRNA JO452541.2 - 2 - 3 272 67

View Full Product Details