Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKRF
Entrez Gene ID
102173820
Gene Name
NFKB repressing factor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189785: 142630 - 156779 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044740.1 XP_017900229.1 2 - 3 448 83

View Full Product Details