Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXB1
Entrez Gene ID
102170178
Gene Name
homeobox B1
Gene Aliases
-
Chromosome Location
Chr.19: 37592070 - 37593509 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064938.1 XP_017920427.1 1 - 2 587 102

View Full Product Details