Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAA30
Entrez Gene ID
102173940
Gene Name
N(alpha)-acetyltransferase 30, NatC catalytic subunit
Gene Aliases
-
Chromosome Location
Chr.10: 33468158 - 33488971 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054097.1 XP_017909586.1 2 - 3 919 111
GenBank mRNA KA339984.1 - 1 - 2 640 111

View Full Product Details