Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNIH3
Entrez Gene ID
102187151
Gene Name
cornichon family AMPA receptor auxiliary protein 3
Gene Aliases
-
Chromosome Location
Chr.16: 26233682 - 26368157 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690521.3 XP_005690578.1 2 - 3 1211 72

View Full Product Details