Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR5
Entrez Gene ID
102175250
Gene Name
phospholipid scramblase family member 5
Gene Aliases
-
Chromosome Location
Chr.1: 121470815 - 121492073 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018049157.1 XP_017904646.1 5 - 6 754 86

View Full Product Details