Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELSPBP1
Entrez Gene ID
102181623
Gene Name
epididymal sperm binding protein 1
Gene Aliases
-
Chromosome Location
Chr.18: 55874532 - 55886769 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692673.3 XP_005692730.1 3 - 4 352 69
GenBank mRNA JO438934.2 - 2 - 3 367 69

View Full Product Details