Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO40
Entrez Gene ID
102180797
Gene Name
F-box protein 40
Gene Aliases
-
Chromosome Location
Chr.1: 65766950 - 65787075 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044230.1 XP_017899719.1 5 - 6 2446 70
GenBank mRNA JO368124.2 - 1 - 2 1726 70
JO368125.2 - 1 - 2 1726 70
JO368126.2 - 1 - 2 404 70
JO377024.2 - 1 - 2 1749 70
JO377025.2 - 1 - 2 1749 70
JO452474.2 - 1 - 2 396 70
JO452530.2 - 1 - 2 395 70
JO556784.2 - 1 - 2 458 70
JO556786.2 - 1 - 2 458 70
JT737675.1 - 2 - 3 1931 70

View Full Product Details