Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA2
Entrez Gene ID
102184431
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
Chromosome Location
Chr.8: 74354144 - 74365049 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684036.3 XP_005684093.2 1 - 2 194 95

View Full Product Details