Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC58
Entrez Gene ID
102171379
Gene Name
leucine rich repeat containing 58
Gene Aliases
-
Chromosome Location
Chr.1: 64773657 - 64791813 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675231.3 XP_005675288.3 2 - 3 734 80
GenBank mRNA JO327471.2 - 2 - 3 672 80
JO362846.2 - 2 - 3 302 80
JO415806.2 - 2 - 3 510 80
JO415807.2 - 2 - 3 510 80
JO430719.2 - 2 - 3 700 80
JO507354.2 - 2 - 3 529 80
JO583963.1 - 2 - 3 670 80
JO583964.1 - 2 - 3 670 80
JO641085.2 - 2 - 3 1094 80
JO641086.2 - 2 - 3 1094 80
JT737667.1 - 2 - 3 496 80
JT762742.1 - 2 - 3 1070 80
KA339544.1 - 2 - 3 1075 80

View Full Product Details