Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNV1
Entrez Gene ID
102185673
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Gene Aliases
-
Chromosome Location
Chr.14: 27730990 - 27738209 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058361.1 XP_017913850.1 3 - 4 2046 60

View Full Product Details