Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRDM14
Entrez Gene ID
102169199
Gene Name
PR/SET domain 14
Gene Aliases
-
Chromosome Location
Chr.14: 48319168 - 48337684 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058124.1 XP_017913613.1 3 - 4 906 62

View Full Product Details