Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMSY
Entrez Gene ID
102175878
Gene Name
EMSY transcriptional repressor, BRCA2 interacting
Gene Aliases
-
Chromosome Location
Chr.15: 26990509 - 27072511 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690007.3 XP_005690064.1 15 - 16 2635 107
XM_005690008.3 XP_005690065.1 15 - 16 2634 107
XM_013969807.2 XP_013825261.1 14 - 15 2593 107
XM_013969808.2 XP_013825262.1 14 - 15 2482 107
XM_013969810.2 XP_013825264.1 14 - 15 2481 107
XM_013969812.2 XP_013825266.1 15 - 16 2636 107
XM_018059320.1 XP_017914809.1 14 - 15 2535 107

View Full Product Details