Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC23
Entrez Gene ID
102187264
Gene Name
zinc finger DHHC-type containing 23
Gene Aliases
-
Chromosome Location
Chr.1: 58387275 - 58399787 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039430.1 XP_017894919.1 3 - 4 1154 82
GenBank mRNA JO338812.2 - 1 - 2 132 82
JO363832.2 - 1 - 2 115 82
JO434432.2 - 3 - 4 944 82
JO591579.2 - 3 - 4 1071 82
JT705700.1 - 4 - 5 1022 82
JT738702.1 - 6 - 7 1049 82
KA353112.1 - 3 - 4 977 82

View Full Product Details