Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD6
Entrez Gene ID
102183834
Gene Name
tudor domain containing 6
Gene Aliases
-
Chromosome Location
Chr.23: 28957069 - 28968463 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039323.1 XP_017894812.1 1 - 2 6056 106
XM_018039324.1 XP_017894813.1 1 - 2 6056 106

View Full Product Details