Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYSMD3
Entrez Gene ID
102178904
Gene Name
LysM domain containing 3
Gene Aliases
-
Chromosome Location
Chr.7: 20444136 - 20457088 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050045.1 XP_017905534.1 2 - 3 493 88
GenBank mRNA JO593089.2 - 1 - 2 957 88
JT660089.1 - 1 - 2 676 88

View Full Product Details