Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMELX
Entrez Gene ID
100860759
Gene Name
amelogenin X-linked
Gene Aliases
AML-X
Chromosome Location
Chr.X|NW_017189516: 58861147 - 58868851 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285616.1 NP_001272545.1 5 - 6 646 70
GenBank mRNA AF215889.1 - 5 - 6 647 70

View Full Product Details