Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO1
Entrez Gene ID
102183890
Gene Name
LIM domain only 1
Gene Aliases
-
Chromosome Location
Chr.15: 38063348 - 38104898 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059499.1 XP_017914988.1 3 - 4 973 89
XM_018059500.1 XP_017914989.1 3 - 4 527 89
GenBank mRNA JT743423.1 - 3 - 4 376 89

View Full Product Details