Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMADHC
Entrez Gene ID
100860757
Gene Name
metabolism of cobalamin associated D
Gene Aliases
-
Chromosome Location
Chr.2: 89704272 - 89721539 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285614.1 NP_001272543.1 2 - 3 370 124
XM_013967881.2 XP_013823335.1 4 - 5 562 124
GenBank mRNA EF151288.1 - 2 - 3 370 124
KA307491.1 - 4 - 5 556 124

View Full Product Details