Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2K
Entrez Gene ID
100860797
Gene Name
ubiquitin conjugating enzyme E2 K
Gene Aliases
HIP2
Chromosome Location
Chr.6: 59364109 - 59433006 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285657.1 NP_001272586.1 5 - 6 402 75
GenBank mRNA EU244429.1 - 5 - 6 402 75
JO393062.2 - 2 - 3 387 75
KA343267.1 - 2 - 3 401 75

View Full Product Details