Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OAZ2
Entrez Gene ID
102169013
Gene Name
ornithine decarboxylase antizyme 2
Gene Aliases
-
Chromosome Location
Chr.10: 57670486 - 57683194 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290120.1 NP_001277049.1 2 - 3 382 85
GenBank mRNA JO318199.2 - 2 - 3 337 85
JO631734.2 - 2 - 3 382 85
KA314417.1 - 2 - 3 481 85

View Full Product Details