The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285570.1 | NP_001272499.1 | 2 - 3 | 196 | 82 |
XM_013965265.2 | XP_013820719.1 | 2 - 3 | 219 | 82 | |
GenBank mRNA | DQ010419.1 | - | 2 - 3 | 196 | 82 |