Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLIN3
Entrez Gene ID
100861244
Gene Name
perilipin 3
Gene Aliases
-
Chromosome Location
Chr.7: 91234310 - 91255710 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285595.1 NP_001272524.1 5 - 6 725 70
XM_018049969.1 XP_017905458.1 5 - 6 755 70
GenBank mRNA HQ846826.1 - 5 - 6 725 70

View Full Product Details