Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNRHR
Entrez Gene ID
100860755
Gene Name
gonadotropin releasing hormone receptor
Gene Aliases
-
Chromosome Location
Chr.6: 84144225 - 84161853 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285612.1 NP_001272541.1 1 - 2 524 70
GenBank mRNA EF150356.1 - 1 - 2 524 70

View Full Product Details