Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OGN
Entrez Gene ID
100860756
Gene Name
osteoglycin
Gene Aliases
-
Chromosome Location
Chr.8: 83789288 - 83805336 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285613.1 NP_001272542.1 1 - 2 170 122
XM_013966068.2 XP_013821522.2 2 - 3 736 122
XM_018051762.1 XP_017907251.1 2 - 3 444 122
GenBank mRNA EF151287.1 - 1 - 2 170 122

View Full Product Details