Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTN1A1
Entrez Gene ID
100860762
Gene Name
butyrophilin subfamily 1 member A1
Gene Aliases
-
Chromosome Location
Chr.23: 18384019 - 18391212 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285618.1 NP_001272547.1 2 - 3 474 86
XM_013974033.2 XP_013829487.1 3 - 4 593 86
XM_013974034.2 XP_013829488.1 3 - 4 603 86
XM_013974035.2 XP_013829489.1 2 - 3 444 86
XM_013974036.2 XP_013829490.1 3 - 4 593 86
GenBank mRNA EF102891.1 - 2 - 3 474 86

View Full Product Details