Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RARRES2
Entrez Gene ID
102190100
Gene Name
retinoic acid receptor responder 2
Gene Aliases
chemerin
Chromosome Location
Chr.4: 7917257 - 7921039 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312693.1 NP_001299622.1 3 - 4 392 65
XM_013963442.2 XP_013818896.1 4 - 5 510 65
XM_018046622.1 XP_017902111.1 4 - 5 524 65
XM_018046623.1 XP_017902112.1 4 - 5 424 65
GenBank mRNA KT313029.1 - 3 - 4 392 65

View Full Product Details