Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASR
Entrez Gene ID
102185783
Gene Name
calcium sensing receptor
Gene Aliases
-
Chromosome Location
Chr.1: 66373772 - 66404333 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286092.1 NP_001273021.1 4 - 5 1805 98
GenBank mRNA JO425752.1 - 4 - 5 1587 98
JO428925.2 - 4 - 5 1527 98
KF006348.1 - 4 - 5 1805 98

View Full Product Details