Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
GRINA
Entrez Gene ID
102181056
Gene Name
glutamate ionotropic receptor NMDA type subunit associated protein 1
Gene Aliases
-
Chromosome Location
Chr.14: 81113503 - 81116799 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058698.1 XP_017914187.1 2 - 3 551 100
GenBank mRNA JO363369.2 - 2 - 3 549 100
JO431649.2 - 2 - 3 536 100

View Full Product Details