Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NECTIN3
Entrez Gene ID
102173039
Gene Name
nectin cell adhesion molecule 3
Gene Aliases
-
Chromosome Location
Chr.1: 55700391 - 55822056 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064281.1 XP_017919770.1 5 - 6 1223 70

View Full Product Details