Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZSCAN20
Entrez Gene ID
102187735
Gene Name
zinc finger and SCAN domain containing 20
Gene Aliases
-
Chromosome Location
Chr.3: 8471953 - 8493933 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678735.3 XP_005678792.2 6 - 7 1567 102
XM_013963012.2 XP_013818466.2 6 - 7 1638 102
XM_018041131.1 XP_017896620.1 6 - 7 1553 102

View Full Product Details