Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPHX3
Entrez Gene ID
102184515
Gene Name
epoxide hydrolase 3
Gene Aliases
-
Chromosome Location
Chr.7: 100801869 - 100816661 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051295.1 XP_017906784.1 1 - 2 1190 66
XM_018051296.1 XP_017906785.1 2 - 3 346 66
XM_018051297.1 XP_017906786.1 1 - 2 1190 66
XM_018051298.1 XP_017906787.1 1 - 2 1190 66
GenBank mRNA JO617569.2 - 1 - 2 1249 66

View Full Product Details