Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GALT
Entrez Gene ID
102172896
Gene Name
galactose-1-phosphate uridylyltransferase
Gene Aliases
-
Chromosome Location
Chr.8: 75837854 - 75841220 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684085.3 XP_005684142.1 7 - 8 785 104
XM_005684087.3 XP_005684144.1 7 - 8 783 104
XM_013966145.2 XP_013821599.1 7 - 8 807 104

View Full Product Details