Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C17H12orf43
Entrez Gene ID
102175507
Gene Name
chromosome 17 C12orf43 homolog
Gene Aliases
-
Chromosome Location
Chr.17: 7927788 - 7938022 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691577.3 XP_005691634.2 2 - 3 212 88
XM_005691578.3 XP_005691635.2 2 - 3 208 88

View Full Product Details