Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSM3
Entrez Gene ID
102190828
Gene Name
LSM3 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
-
Chromosome Location
Chr.22: 57937794 - 57976033 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696141.3 XP_005696198.1 3 - 4 315 105

View Full Product Details